Anti-Kinesin 5A Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB5628-100EA 560.23 CHF
ABCAAB5628-100
Anti-Kinesin 5A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Kinesin 5A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR