Anti-PKC beta 1 + PKC beta 2 Rabbit polyclonal antibody

ABCAAB5627-100EA 708 CHF
ABCAAB5627-100
Anti-PKC beta 1 + PKC beta 2 Rabbit polyclonal antibody
Type: Primary
Antigen: PKC beta 1 + PKC beta 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR