Anti-THRB Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB5622-100EA 534.19 CHF
ABCAAB5622-100
Anti-THRB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: THRB (thyroid hormone receptor, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR