Anti-IRS1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB5599-100EA 492.8 CHF
ABCAAB5599-100
Anti-IRS1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IRS1 (insulin receptor substrate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR