Anti-CRYAB Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB5597-100EA 924 CHF
ABCAAB5597-100
Anti-CRYAB Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR