Anti-CALM1 Mouse monoclonal antibody [clone: 6D4]

ABCAAB5494-200EA 708 CHF
ABCAAB5494-200
Anti-CALM1 Mouse monoclonal antibody [clone: 6D4]
Type: Primary
Antigen: CALM1 (calmodulin 1 (phosphorylase kinase, delta))
Clonality: monoclonal
Clone: 6D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR