Anti-RAC1 + Cdc42 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB5482-50EA 518.74 CHF
ABCAAB5482-50
Anti-RAC1 + Cdc42 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAC1 + Cdc42
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR