Anti-KDR Rabbit polyclonal antibody

ABCAAB5464-50EA 660 CHF
ABCAAB5464-50
Anti-KDR Rabbit polyclonal antibody
Type: Primary
Antigen: KDR (kinase insert domain receptor (a type III receptor tyrosine kinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR