Anti-PI 3 Kinase regulatory subunit 4 Rabbit polyclonal antibody

ABCAAB5461-100EA 672 CHF
ABCAAB5461-100
Anti-PI 3 Kinase regulatory subunit 4 Rabbit polyclonal antibody
Type: Primary
Antigen: PI 3 Kinase regulatory subunit 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR