Anti-Blooms Syndrome Protein Blm Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB5446-50EA 492.8 CHF
ABCAAB5446-50
Anti-Blooms Syndrome Protein Blm Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Blooms Syndrome Protein Blm
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR