Anti-CAPN1 Mouse monoclonal antibody [clone: 2H2A7C2]

ABCAAB5441-100EA 725 CHF
ABCAAB5441-100
Anti-CAPN1 Mouse monoclonal antibody [clone: 2H2A7C2]
Type: Primary
Antigen: CAPN1 (calpain 1, (mu/I) large subunit)
Clonality: monoclonal
Clone: 2H2A7C2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR