Anti-HSPA13 Mouse Monoclonal Antibody [clone: 3A3]

Supplier: Abcam
ABCAAB5439-50EA 979 CHF
ABCAAB5439-50
Anti-HSPA13 Mouse Monoclonal Antibody [clone: 3A3]
Antibodies
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: monoclonal
Clone: 3A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR