Anti-MEK7 Rabbit polyclonal antibody

ABCAAB5436-100EA 672 CHF
ABCAAB5436-100
Anti-MEK7 Rabbit polyclonal antibody
Type: Primary
Antigen: MEK7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR