Anti-Retinoic Acid Receptor alpha Mouse monoclonal antibody unconjugated [clone: Ralpha10]

ABCAAB5423-50EA 708 CHF
ABCAAB5423-50
Anti-Retinoic Acid Receptor alpha Mouse monoclonal antibody unconjugated [clone: Ralpha10]
Type: Primary
Antigen: Retinoic Acid Receptor alpha
Clonality: monoclonal
Clone: Ralpha10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR