Anti-FANCD2 Mouse monoclonal antibody [clone: FI-17]

ABCAAB5360-100EA 732 CHF
ABCAAB5360-100
Anti-FANCD2 Mouse monoclonal antibody [clone: FI-17]
Type: Primary
Antigen: FANCD2 (Fanconi anemia, complementation group D2)
Clonality: monoclonal
Clone: FI-17
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR