Anti-PPP1R8 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB5300-100EA 508.36 CHF
ABCAAB5300-100
Anti-PPP1R8 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP1R8 (protein phosphatase 1, regulatory subunit 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR