Anti-MARK4 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB5262-100EA 472.05 CHF
ABCAAB5262-100
Anti-MARK4 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MARK4 (MAP/microtubule affinity-regulating kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR