Anti-PAK1 + PAK2 + PAK3 Rabbit polyclonal antibody

Supplier: Abcam
ABCAAB5247-50EA 508.36 CHF
ABCAAB5247-50
Anti-PAK1 + PAK2 + PAK3 Rabbit polyclonal antibody
Type: Primary
Antigen: PAK1 + PAK2 + PAK3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR