Anti-IFT57 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB5205-100EA 497.99 CHF
ABCAAB5205-100
Anti-IFT57 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IFT57 (intraflagellar transport 57 homolog (Chlamydomonas))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR