Anti-BRDT Rabbit polyclonal antibody

ABCAAB5157-100EA 691 CHF
ABCAAB5157-100
Anti-BRDT Rabbit polyclonal antibody
Type: Primary
Antigen: BRDT (bromodomain, testis-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR