Anti-AGO2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB5072-100EA 451.3 CHF
ABCAAB5072-100
Anti-AGO2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AGO2 (argonaute RISC catalytic component 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR