Anti-LCK Rabbit polyclonal antibody

ABCAAB4900-50EA 744 CHF
ABCAAB4900-50
Anti-LCK Rabbit polyclonal antibody
Type: Primary
Antigen: LCK (lymphocyte-specific protein tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR