Anti-IRS1 Rabbit polyclonal antibody

Supplier: Abcam
ABCAAB4873-50EA 508.36 CHF
ABCAAB4873-50
Anti-IRS1 Rabbit polyclonal antibody
Type: Primary
Antigen: IRS1 (insulin receptor substrate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR