Anti-PXN Rabbit polyclonal antibody

Supplier: Abcam
ABCAAB4833-50EA 518.74 CHF
ABCAAB4833-50
Anti-PXN Rabbit polyclonal antibody
Type: Primary
Antigen: PXN (paxillin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR