Anti-CDK1 Rabbit polyclonal antibody

ABCAAB4823-50EA 660 CHF
ABCAAB4823-50
Anti-CDK1 Rabbit polyclonal antibody
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR