Anti-RANGAP1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB4784-200EA 945 CHF
ABCAAB4784-200
Anti-RANGAP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RANGAP1 (Ran GTPase activating protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR