Anti-ATG12 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB4752-250EA 508.36 CHF
ABCAAB4752-250
Anti-ATG12 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: ATG12 (autophagy related 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR