Anti-SMARCAL1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB4743-100EA 870 CHF
ABCAAB4743-100
Anti-SMARCAL1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SMARCAL1 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR