Anti-HIST4H4 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB4627-100EA 518.74 CHF
ABCAAB4627-100
Anti-HIST4H4 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR