Anti-C11orf30 Rabbit polyclonal antibody

ABCAAB4580-200EA 660 CHF
ABCAAB4580-200
Anti-C11orf30 Rabbit polyclonal antibody
Type: Primary
Antigen: C11orf30 (chromosome 11 open reading frame 30)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR