Anti-FBL Mouse monoclonal antibody unconjugated [clone: 28F2]

Lieferant: Abcam
ABCAAB4575-250EA 903 CHF
ABCAAB4575-250
Anti-FBL Mouse monoclonal antibody unconjugated [clone: 28F2]
Antikörper
Type: Primary
Antigen: FBL (fibrillarin)
Clonality: monoclonal
Clone: 28F2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR