Anti-CEACAM 5 + 6 Mouse monoclonal antibody unconjugated [clone: MUS]

Supplier: Abcam
ABCAAB4539-50EA 472.05 CHF
ABCAAB4539-50
Anti-CEACAM 5 + 6 Mouse monoclonal antibody unconjugated [clone: MUS]
Antibodies
Type: Primary
Antigen: CEACAM 5 + 6
Clonality: monoclonal
Clone: MUS
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR