Anti-MORF4L1 Rabbit polyclonal antibody

ABCAAB4467-100EA 672 CHF
ABCAAB4467-100
Anti-MORF4L1 Rabbit polyclonal antibody
Type: Primary
Antigen: MORF4L1 (mortality factor 4 like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR