Anti-SA2 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB4463-50EA 539.48 CHF
ABCAAB4463-50
Anti-SA2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SA2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR