Anti-MCM4 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB4459-50EA 529.11 CHF
ABCAAB4459-50
Anti-MCM4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MCM4 (minichromosome maintenance complex component 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR