Anti-TRIM3 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB4215-100EA 881 CHF
ABCAAB4215-100
Anti-TRIM3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRIM3 (tripartite motif containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR