Anti-SFRP1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB4193-100EA 0 CHF
ABCAAB4193-100
Anti-SFRP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SFRP1 (secreted frizzled-related protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR