Anti-PRKCG Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB4145-100EA 911 CHF
ABCAAB4145-100
Anti-PRKCG Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRKCG (protein kinase C, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR