Anti-PRKCZ Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB4137-100EA 933 CHF
ABCAAB4137-100
Anti-PRKCZ Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRKCZ (protein kinase C, zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR