Anti-GABRA4 Rabbit polyclonal antibody

ABCAAB4120-100EA 762 CHF
ABCAAB4120-100
Anti-GABRA4 Rabbit polyclonal antibody
Type: Primary
Antigen: GABRA4 (gamma-aminobutyric acid (GABA) A receptor, alpha 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR