Anti-HIST3H3 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB4076-100EA 534.3 CHF
ABCAAB4076-100
Anti-HIST3H3 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR