Anti-ALK Rabbit polyclonal antibody

ABCAAB4061-500EA 660 CHF
ABCAAB4061-500
Anti-ALK Rabbit polyclonal antibody
Type: Primary
Antigen: ALK (anaplastic lymphoma receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR