Anti-RFPL1 Goat polyclonal antibody unconjugated

ABCAAB4002-100EA 441 CHF
ABCAAB4002-100
Anti-RFPL1 Goat polyclonal antibody unconjugated
Type: Primary
Antigen: RFPL1 (ret finger protein-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR