Anti-Blood Group Lewis b Mouse Monoclonal Antibody [clone: 2-25LE]

Supplier: Abcam
ABCAAB3968-100EA 508.36 CHF
ABCAAB3968-100
Anti-Blood Group Lewis b Mouse Monoclonal Antibody [clone: 2-25LE]
Antibodies
Type: Primary
Antigen: Blood Group Lewis b
Clonality: monoclonal
Clone: 2-25LE
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR