Anti-H3F3AP6 Mouse monoclonal antibody [clone: DCS-60.2]

ABCAAB3929-500EA 744 CHF
ABCAAB3929-500
Anti-H3F3AP6 Mouse monoclonal antibody [clone: DCS-60.2]
Type: Primary
Antigen: H3F3AP6 (H3 histone, family 3A, pseudogene 6)
Clonality: monoclonal
Clone: DCS-60.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR