Anti-DOPA Decarboxylase Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB3905-100EA 577.12 CHF
ABCAAB3905-100
Anti-DOPA Decarboxylase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DOPA Decarboxylase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat