Anti-GRIN2B Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3856-100EA 508.36 CHF
ABCAAB3856-100
Anti-GRIN2B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GRIN2B (glutamate receptor, ionotropic, N-methyl D-aspartate 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR