Anti-HELLS Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3851-50EA 891 CHF
ABCAAB3851-50
Anti-HELLS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HELLS (helicase, lymphoid-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR