Anti-DCLRE1C Goat polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB3834-100EA 911 CHF
ABCAAB3834-100
Anti-DCLRE1C Goat polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: DCLRE1C (DNA cross-link repair 1C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR