Anti-SETMAR Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB3823-100EA 440.92 CHF
ABCAAB3823-100
Anti-SETMAR Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SETMAR (SET domain and mariner transposase fusion gene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR